ContactUsLinkComponent
White, low profile, pcr 8-tube strips with individually hinged caps, 0.1ml, case of 120. op 1 * 120 items
Catalog # GREI671221_120
Supplier:  Greiner Bio-One
White, low profile, pcr 8-tube strips with individually hinged caps, 0.1ml, case of 120. op 1 * 120 items
Catalog # GREI671221_120
Supplier:  Greiner Bio-One
Frequently Bought Together