ContactUsLinkComponent
Cogent HPS C18 HPLC Column 120A 5um High Purity Silica. 21.2mm ID x 150mm. 1 * 1 items
Catalog # MICS75018-P150
Supplier:  MICROSOLV
undefined
Cogent HPS C18 HPLC Column 120A 5um High Purity Silica. 21.2mm ID x 150mm. 1 * 1 items
Catalog # MICS75018-P150
Supplier:  MICROSOLV
Frequently Bought Together