ContactUsLinkComponent
Capillary, Controlled Flow, high flow 75um ID by 1 meter. Use in Capillary Electrophoresis. 1/pack. 1 * 1 m
Catalog # MICS04675-HF
Supplier:  MICROSOLV
undefined
Capillary, Controlled Flow, high flow 75um ID by 1 meter. Use in Capillary Electrophoresis. 1/pack. 1 * 1 m
Catalog # MICS04675-HF
Supplier:  MICROSOLV
Frequently Bought Together