ContactUsLinkComponent
Plunger Dispensing Can, 2 quart, perforated pan screen serves as flame arrester, Steel, Yellow. 1 * 1 items
Catalog # 119-0264
Plunger Dispensing Can, 2 quart, perforated pan screen serves as flame arrester, Steel, Yellow. 1 * 1 items
Catalog # 119-0264
Frequently Bought Together