ContactUsLinkComponent
Electrode holder With cable duct. For three electrodes, opening O 16 mm. Additional opening O 11 mm. Three joints for high flexibility and adjustment options. Base plate: 110 x 130 mm. Base plate weight: 0.6 kg. Arm weight: 0.1 kg. Max. arm height: 3 1 * 1 items
Catalog # ROTHEHY1.1
Supplier:  Roth Carl
Electrode holder With cable duct. For three electrodes, opening O 16 mm. Additional opening O 11 mm. Three joints for high flexibility and adjustment options. Base plate: 110 x 130 mm. Base plate weight: 0.6 kg. Arm weight: 0.1 kg. Max. arm height: 3 1 * 1 items
Catalog # ROTHEHY1.1
Supplier:  Roth Carl
Frequently Bought Together