ContactUsLinkComponent
THERMOSTATIC REACTION VESSEL, 2000 ML, DN 120, FLAT FLANGE WITH GROOVE, WITH 2 SCREW THREADS GL 18, FLAT BOTTOM, UNGRADUATED, OUTER Ø: 150 MM, INNER Ø: 120 MM, HEIGHT: 275 MM BOROSILICATE GLASS 1 * 1 items
Catalog # WITG0.992.571
Supplier:  witeg Labortechnik
THERMOSTATIC REACTION VESSEL, 2000 ML, DN 120, FLAT FLANGE WITH GROOVE, WITH 2 SCREW THREADS GL 18, FLAT BOTTOM, UNGRADUATED, OUTER Ø: 150 MM, INNER Ø: 120 MM, HEIGHT: 275 MM BOROSILICATE GLASS 1 * 1 items
Catalog # WITG0.992.571
Supplier:  witeg Labortechnik
Frequently Bought Together