Dutasteride 99%
Glass bottle, 250 mg
Catalog # ACRO461512500
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:164656-23-9
- Formula:C₂₇H₃₀F₆N₂O₂
- MDL Number:MFCD00937869
- Molecular Weight:528.54 g/mol
- Melting Pt:245 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 %
- HPLC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:250 mg



