Dutasteride 99%
Glass bottle, 1 g
Catalog # ACRO461510010
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:164656-23-9
- Formula:C₂₇H₃₀F₆N₂O₂
- MDL Number:MFCD00937869
- Molecular Weight:528.54 g/mol
- Melting Pt:245 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 %
- HPLC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



