ContactUsLinkComponent
Slide CELLview 75x25mm polystyrene glass bottom Advanced tissue culture surface 10 compartments sterile (5 pieces/bag) 1 * 45 items
Catalog # GREI543979_45
Supplier:  Greiner Bio-One
Slide CELLview 75x25mm polystyrene glass bottom Advanced tissue culture surface 10 compartments sterile (5 pieces/bag) 1 * 45 items
Catalog # GREI543979_45
Supplier:  Greiner Bio-One
Frequently Bought Together