£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
Tube Vacuette 1ml 13x75mm K2E K2EDTA lavender cap-white ring sterile 1 * 1.200 items
GREI454052_1200
:
Greiner Bio-One
Tube Vacuette 1ml 13x75mm K2E K2EDTA lavender cap-white ring sterile 1 * 1.200 items
GREI454052_1200
:
Greiner Bio-One
Frequently Bought Together
Nitrile gloves, TouchNTuff® 92-600 / 92-605
: Ansell
VWR®, Microscope Slides
: VWR Collection
Sterican® Single-Use Needles for Special Applications
: B. Braun
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
: Merck
Water, Ultrapur for trace analysis, Supelco®
: Merck
Test tubes
: Greiner Bio-One
Single use syringes, 2-piece, Injekt® Solo
: B. Braun
Fine dosage syringes, Injekt®-F Solo
: B. Braun
Single-use syringes, 2-piece, HENKE-JECT®
: HENKE SASS WOLF
Single-use syringes, 3-piece, HENKE-JECT®
: HENKE SASS WOLF