THE PERKINELMER 90 MM PLASTIC PETRI DISHES ARE SUITABLE FOR NIRA II 100 MM SPINNER. INCLUDES PACK OF 600 TRANSPARENT POLYSTYRENE PET RI DISHES WITH LIDS 90 MM DIAMETER. 1 * 1 items
Catalog # PERCL9004575
Supplier: PerkinElmer
THE PERKINELMER 90 MM PLASTIC PETRI DISHES ARE SUITABLE FOR NIRA II 100 MM SPINNER. INCLUDES PACK OF 600 TRANSPARENT POLYSTYRENE PET RI DISHES WITH LIDS 90 MM DIAMETER. 1 * 1 items
Catalog # PERCL9004575
Supplier: PerkinElmer
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Screw Cap, 5,0 ml
Supplier: EPPENDORF
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Exhaust filters, V3.0
Supplier: S.C.A.T.
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals