ContactUsLinkComponent
CALIBRATED NOMINALLY 50% REFLECTIVE SINTERED PTFE POLYMER DISC 8 MM THICK X 98 MM DIAMETER WITHIN A SEALABLE CONTAINER SUITABLE FOR THE NIRA II 100 MM SPINNER. 1 * 1 items
Catalog # PERCL1281925
Supplier:  PerkinElmer
CALIBRATED NOMINALLY 50% REFLECTIVE SINTERED PTFE POLYMER DISC 8 MM THICK X 98 MM DIAMETER WITHIN A SEALABLE CONTAINER SUITABLE FOR THE NIRA II 100 MM SPINNER. 1 * 1 items
Catalog # PERCL1281925
Supplier:  PerkinElmer
Frequently Bought Together