BIOLAMINA BIOLAMININ 521 LN 1 x 5 ml 1 * 500 µG
Catalog # MSPPLN521-05
Supplier: MARKET SOURCE PART PROCESS
BIOLAMINA BIOLAMININ 521 LN 1 x 5 ml 1 * 500 µG
Catalog # MSPPLN521-05
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
VWR®, Ice Pans, with or without Lid
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Screw Cap, 5,0 ml
Supplier: EPPENDORF
Corning® CoolCell® containers
Supplier: Corning
CATHEPSIN B ASSAY KIT MAGIC RED 1 * 100 Tests
Supplier: Abcam
EDTA disodium salt dihydrate ≥99% for biotechnology
Supplier: VWR Chemicals
Anti-Glucagon Mouse Monoclonal Antibody [clone: K79bB10]
Supplier: Abcam
Square cover slips
Supplier: MARIENFELD