200MM STAINLESS STEEL FRAME AND MESH SIEVE 63UM APERTURE SIZE 100MM HEIGHT TO ISO3310/1 1 * 1 items
ENDE200SIW.063X100
200MM STAINLESS STEEL FRAME AND MESH SIEVE 63UM APERTURE SIZE 100MM HEIGHT TO ISO3310/1 1 * 1 items
ENDE200SIW.063X100
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection