ContactUsLinkComponent
tris-acetate sds buffer kit is a complete kit to resolve high molecular weight proteins (36-400 kda) under denaturing conditions on tris/acetate gels. 1 * 1 KIT
  GENO786-589
tris-acetate sds buffer kit is a complete kit to resolve high molecular weight proteins (36-400 kda) under denaturing conditions on tris/acetate gels. 1 * 1 KIT
  GENO786-589
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.