ContactUsLinkComponent
protein purification: g-biosciences, spin column, 5ml these empty, plastic, disposable 5ml spin columns have an internal volume of 8 .5ml and is designed for small scale purifications using 15ml centrifuge tubes. 1 * 25 items
Catalog # GENO786-981
Supplier:  G-Biosciences
protein purification: g-biosciences, spin column, 5ml these empty, plastic, disposable 5ml spin columns have an internal volume of 8 .5ml and is designed for small scale purifications using 15ml centrifuge tubes. 1 * 25 items
Catalog # GENO786-981
Supplier:  G-Biosciences
Frequently Bought Together