ContactUsLinkComponent
protein purification: g-biosciences, spin column, 10ml these empty, plastic, disposable 10ml spin columns have an internal volume of 22ml and is designed for small scale purifications using 50ml centrifuge tubes. 1 * 25 items
Catalog # GENO786-983
Supplier:  G-Biosciences
protein purification: g-biosciences, spin column, 10ml these empty, plastic, disposable 10ml spin columns have an internal volume of 22ml and is designed for small scale purifications using 50ml centrifuge tubes. 1 * 25 items
Catalog # GENO786-983
Supplier:  G-Biosciences
Frequently Bought Together