ContactUsLinkComponent
nhs-activated agarose consists of 4% cross-linked agarose that has been activated by the addition of a reactive nhs (n-hydroxysuccin imide) group. the nhs group forms covalent, chemically stable amide bonds with ligands that contain primary amines. 1 * 1 g
Catalog # GENO786-1211
Supplier:  G-Biosciences
nhs-activated agarose consists of 4% cross-linked agarose that has been activated by the addition of a reactive nhs (n-hydroxysuccin imide) group. the nhs group forms covalent, chemically stable amide bonds with ligands that contain primary amines. 1 * 1 g
Catalog # GENO786-1211
Supplier:  G-Biosciences