ContactUsLinkComponent
columns for separation of biomolecules via hydrophobic interaction chromatography (hic). biomolecules are separated based on hydroph obicity of ligand attached to the resin, the distribution of surface-exposed hydrophobic amino acids present on the p 1 * 5 mL
Catalog # GENO786-1002
Supplier:  G-Biosciences
columns for separation of biomolecules via hydrophobic interaction chromatography (hic). biomolecules are separated based on hydroph obicity of ligand attached to the resin, the distribution of surface-exposed hydrophobic amino acids present on the p 1 * 5 mL
Catalog # GENO786-1002
Supplier:  G-Biosciences