5 components Chlorides (Cl-) 0.05mg/l Nitrites (NO2-) 0.05mg/l Nitrates (NO3-) 0.05mg/l Phosphates (PO43-) 0.05mg/l Sulphates (SO42-) 0.1mg/l in Water . Duree de conservation : 4 mois a ouverture du flacon, Duree de validite: 8 mois. 1 * 100 mL
Catalog # CMPLFDC2.D05.W.L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together