ContactUsLinkComponent
Tubing threaded 12x110x1.5mm rd14 with screw threaded straight,acc. to din 12216, with iso screw thread,made of borosilicate glass, available with screw thread gl, rd or svl null
Catalog # WITG0.182.214
Supplier:  witeg Labortechnik
Tubing threaded 12x110x1.5mm rd14 with screw threaded straight,acc. to din 12216, with iso screw thread,made of borosilicate glass, available with screw thread gl, rd or svl null
Catalog # WITG0.182.214
Supplier:  witeg Labortechnik
Frequently Bought Together