ContactUsLinkComponent
Monoclonal anti-GFP is designed to detect enhanced GFP and GFP containing recombinant proteins. This product is prepared by labeling high quality monoclonal mouse anti-GFP IgG (H+L) with the red fluorescent dye CF™568. 1 * 100 µl
Catalog # BTIU20480
Supplier:  Biotium
Monoclonal anti-GFP is designed to detect enhanced GFP and GFP containing recombinant proteins. This product is prepared by labeling high quality monoclonal mouse anti-GFP IgG (H+L) with the red fluorescent dye CF™568. 1 * 100 µl
Catalog # BTIU20480
Supplier:  Biotium
Frequently Bought Together