ContactUsLinkComponent
Beaker Quartz glass in application temperatures of up to 1050C (maximum 1600C), 250 ml volume, wide form, with lip and spout, tr anslucent, high chemical resistance and purity. Diameter 68mm, height 88mm. The durability or operating cycles of our q 1 * 1 items
Catalog # TSLA73161
Beaker Quartz glass in application temperatures of up to 1050C (maximum 1600C), 250 ml volume, wide form, with lip and spout, tr anslucent, high chemical resistance and purity. Diameter 68mm, height 88mm. The durability or operating cycles of our q 1 * 1 items
Catalog # TSLA73161
Frequently Bought Together