ContactUsLinkComponent
Tray Quatrz glass in application temperatures of up to 1050C (maximum 1600C), 3-sided, with open end, translucent, high chemical resistance and purity. Length 114mm, width 67mm, height 25mm. The durability or operating cycles of our quartz glass c 1 * 1 items
Catalog # TSLA73041
Tray Quatrz glass in application temperatures of up to 1050C (maximum 1600C), 3-sided, with open end, translucent, high chemical resistance and purity. Length 114mm, width 67mm, height 25mm. The durability or operating cycles of our quartz glass c 1 * 1 items
Catalog # TSLA73041
Frequently Bought Together