ContactUsLinkComponent
The PTFE Template tool allows a fixed area to be swabbed. A PTFE template designed to a predetermined area ensures swabbing is carried out over a set area. This gives improved results. Hole 10x105 cm. 1 * 1 items
Catalog # 300-0417
Supplier:  Sampling Systems
The PTFE Template tool allows a fixed area to be swabbed. A PTFE template designed to a predetermined area ensures swabbing is carried out over a set area. This gives improved results. Hole 10x105 cm. 1 * 1 items
Catalog # 300-0417
Supplier:  Sampling Systems
Frequently Bought Together