Steroid 5α-Reductase is an important enzyme in androgen physiology because it catalyzes the conversion of testosterone into the more potent 5α-dihydro-testosterone, which mediates androgen effects on target tissues. 1 * 100 µG
Catalog # BWRLBS61336
Supplier: Bioworld Technology