Phosphatidylinositol-glycans (PIGs) are multi-pass transmembrane proteins that localize to the endoplasmic reticulum. PIGs exhibit v arious functions, but all are crucial for the biosynthesis of the glycosylphosphatidylinositol (GPI)-anchor. 1 * 100 µG
Catalog # BWRLBS61315
Supplier: Bioworld Technology