This gene encodes a branched chain aminotransferase found in mitochondria. The encoded protein forms a dimer that catalyzes the firs t step in the production of the branched chain amino acids leucine, isoleucine, and valine. 1 * 100 µG
Catalog # BWRLBS8888
Supplier: Bioworld Technology