KLHL29 (Kelch-like protein 29), also known as KBTBD9, is a 655 amino acid protein that is related to the Drosophila Kelch protein. M utations affecting Kelch function result in failure of Kelch to associate with the ring canals and subsequent female 1 * 100 µG
Catalog # BWRLBS61204
Supplier: Bioworld Technology