The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-tr ans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. 1 * 100 µG
Catalog # BWRLBS8604
Supplier: Bioworld Technology