Thioredoxin 2 (Trx-2) is a small redox protein that is localized to the mitochondria and is essential for cell viability, playing a crucial role in the scavenging of ROS in mitochondria and regulating the mitochondrial apoptosis signaling pathway. 1 * 100 µG
Catalog # BWRLBS8545
Supplier: Bioworld Technology