eEF2K is 105 kDa protein that is detected in skeletal muscle extracts and is phosphorylated rapidly by SAPK4, but poorly by p38, p38 γ, JNK or ERK 2. SAPK4 phosphorylates eEF2K at Ser 359 and Ser 396 in vitro, causing its inactivation. 1 * 100 µG
Catalog # BWRLBS8474
Supplier: Bioworld Technology