Smad proteins, the mammalian homologs of the Drosophila Mothers against dpp (Mad), have been implicated as downstream effectors of T GFß/BMP signaling. Smad1 (also designated Madr1 or JV4-1) and Smad5 are effectors of BMP2 and BMP4 function. 1 * 100 µG
Catalog # BWRLBS64037
Supplier: Bioworld Technology