Troponin, working in conjunction with tropomyosin, functions as a molecular switch that regulates muscle contraction in response to changes in the intracellular Ca2+ concentration. 1 * 100 µG
Catalog # BWRLBS61520
Supplier: Bioworld Technology