The protein encoded by this gene belongs to the CTF/NF-I family. These are dimeric DNA-binding proteins, and function as cellular tr anscription factors and as replication factors for adenovirus DNA replication. 1 * 100 µG
Catalog # BWRLMB0190
Supplier: Bioworld Technology