ContactUsLinkComponent
This product is prepared by labeling goat anti-llama IgG (H+L) with horseradish peroxidase. HRP conjugates could be used for Western Blot, ELISA, immunohistochemistry and other standard immunoassay applications. 1 * 1 mL
  20475-1ML
 :  Biotium
This product is prepared by labeling goat anti-llama IgG (H+L) with horseradish peroxidase. HRP conjugates could be used for Western Blot, ELISA, immunohistochemistry and other standard immunoassay applications. 1 * 1 mL
  20475-1ML
 :  Biotium
Frequently Bought Together