ContactUsLinkComponent
This product is prepared by labeling goat anti-llama IgG (H+L) with horseradish peroxidase. HRP conjugates could be used for Western Blot, ELISA, immunohistochemistry and other standard immunoassay applications. 1 * 100 µl
Catalog # 20475-100UL
Supplier:  Biotium
This product is prepared by labeling goat anti-llama IgG (H+L) with horseradish peroxidase. HRP conjugates could be used for Western Blot, ELISA, immunohistochemistry and other standard immunoassay applications. 1 * 100 µl
Catalog # 20475-100UL
Supplier:  Biotium
Frequently Bought Together