ContactUsLinkComponent
This product is prepared by labeling highly cross-adsorbed goat anti-human IgG (H+L) with horseradish peroxidase. To minimize cross- reactivity, the antibody has been adsorbed against bovine, horse, and mouse serum. HRP conjugates could be used for W 1 * 100 µl
Catalog # 20470-100UL
Supplier:  Biotium
This product is prepared by labeling highly cross-adsorbed goat anti-human IgG (H+L) with horseradish peroxidase. To minimize cross- reactivity, the antibody has been adsorbed against bovine, horse, and mouse serum. HRP conjugates could be used for W 1 * 100 µl
Catalog # 20470-100UL
Supplier:  Biotium