ContactUsLinkComponent
MULTIPURPOSE PFA FAST LOOPS FOR HIGH-FLOW VALVES. RECOMMENDED FOR USE WITH 1.0 MM I.D. (GRAY) PROBE. 1.6 MM O.D., 1/4-28 FITTINGS. 1 * 1 items
Catalog # PERCN0777665
Supplier:  PerkinElmer
MULTIPURPOSE PFA FAST LOOPS FOR HIGH-FLOW VALVES. RECOMMENDED FOR USE WITH 1.0 MM I.D. (GRAY) PROBE. 1.6 MM O.D., 1/4-28 FITTINGS. 1 * 1 items
Catalog # PERCN0777665
Supplier:  PerkinElmer
Frequently Bought Together