ContactUsLinkComponent
USED WITH OPTIONAL ACCESSORY FOR EXHAUSTING TITAN MPS OVEN CAVITY IN LABORATORIES THAT DO NOT HAVE EXHAUST SYSTEMS. EXHAUST TUBE 1.5 M (ID = 76 MM, OD = 82 MM) (ONE INCLUDED WITH TITAN MPS). 1 * 1 items
Catalog # PERCN3134015
Supplier:  PerkinElmer
USED WITH OPTIONAL ACCESSORY FOR EXHAUSTING TITAN MPS OVEN CAVITY IN LABORATORIES THAT DO NOT HAVE EXHAUST SYSTEMS. EXHAUST TUBE 1.5 M (ID = 76 MM, OD = 82 MM) (ONE INCLUDED WITH TITAN MPS). 1 * 1 items
Catalog # PERCN3134015
Supplier:  PerkinElmer
Frequently Bought Together