ContactUsLinkComponent
6MM PFA SECURE NUT FOR ESI CYCLONIC SPRAY CHAMBERS, REVISION B23MM LENGTH WITH 7/16-20 THREADS. SECURES 6MM NEBULIZER TO ESI CHAMBER . 1 * 1 items
Catalog # PERCN8145321
Supplier:  PerkinElmer
6MM PFA SECURE NUT FOR ESI CYCLONIC SPRAY CHAMBERS, REVISION B23MM LENGTH WITH 7/16-20 THREADS. SECURES 6MM NEBULIZER TO ESI CHAMBER . 1 * 1 items
Catalog # PERCN8145321
Supplier:  PerkinElmer
Frequently Bought Together