ContactUsLinkComponent
Goat anti-Mouse IgG (H&L), F(ab)'2 Fragment, min. cross-reactivity to bovine, goat, human, rabbit, or rat IgG (highly absorbed again st rat IgG) 1 * 1 mg
Catalog # AGRIAS163460
Supplier:  Agrisera
Goat anti-Mouse IgG (H&L), F(ab)'2 Fragment, min. cross-reactivity to bovine, goat, human, rabbit, or rat IgG (highly absorbed again st rat IgG) 1 * 1 mg
Catalog # AGRIAS163460
Supplier:  Agrisera