ContactUsLinkComponent
Software easyEmission , incl. USB connection cable for connecting flue gas analyser testo 340 or testo 350 to PC Functions: measurement intervals, readings transferred in seconds to Microsoft EXCEL, user-defined fuels, readings shown in 1 * 1 items
Catalog # TEST0554.3334
Supplier:  Testo
Software easyEmission , incl. USB connection cable for connecting flue gas analyser testo 340 or testo 350 to PC Functions: measurement intervals, readings transferred in seconds to Microsoft EXCEL, user-defined fuels, readings shown in 1 * 1 items
Catalog # TEST0554.3334
Supplier:  Testo
Frequently Bought Together