ContactUsLinkComponent
REAKT.GEF. 250ML DN60 GL14 TEMP.MANTEL DN60 M.RINGNUT, M.PE-OLIVEN, TEMPERIER- BODEN GERADE, BORO 3.3 D. AUßEN 100MM, 180MM HÖHE 1 * 1 items
Catalog # NEUB5020-10-0250
REAKT.GEF. 250ML DN60 GL14 TEMP.MANTEL DN60 M.RINGNUT, M.PE-OLIVEN, TEMPERIER- BODEN GERADE, BORO 3.3 D. AUßEN 100MM, 180MM HÖHE 1 * 1 items
Catalog # NEUB5020-10-0250
Frequently Bought Together