ContactUsLinkComponent
4 – 20 ma current loop interface temperature controller, actual value (0 to +70 °c = 4 – 20 ma) 1 * 1 items
Catalog # MEMMICO150V3
Supplier:  MEMMERT
4 – 20 ma current loop interface temperature controller, actual value (0 to +70 °c = 4 – 20 ma) 1 * 1 items
Catalog # MEMMICO150V3
Supplier:  MEMMERT
Frequently Bought Together