ContactUsLinkComponent
Peltier cooling unit enables a working temperature of 37 c even at higher ambient temperatures of up to 35 c (available as of the fourth quarter of 2016) 1 * 1 items
Catalog # MEMMICO50K5
Supplier:  MEMMERT
Peltier cooling unit enables a working temperature of 37 c even at higher ambient temperatures of up to 35 c (available as of the fourth quarter of 2016) 1 * 1 items
Catalog # MEMMICO50K5
Supplier:  MEMMERT
Frequently Bought Together