ContactUsLinkComponent
Digital Burette Titrex 0-50ml, Type: Standard, Volume: 0-50 ml, Accuracy: ±0.03 m l, Coefficient of variation: ≤0.01 ml, Your advant ages at a glance: high quality precision instrument, made in Germany, highest accuracy and precision, our error range 1 * 1 items
Catalog # WITG5.497.050
Supplier:  witeg Labortechnik
Digital Burette Titrex 0-50ml, Type: Standard, Volume: 0-50 ml, Accuracy: ±0.03 m l, Coefficient of variation: ≤0.01 ml, Your advant ages at a glance: high quality precision instrument, made in Germany, highest accuracy and precision, our error range 1 * 1 items
Catalog # WITG5.497.050
Supplier:  witeg Labortechnik
Frequently Bought Together