Orders LinkContactUsLinkComponent
Methylated DNA IP kit, BIOARRAY™
Methylated DNA IP kit, BIOARRAY™
BIOARRAY™ Methylated DNA IP kit, 10 Tests
Catalog # ALEXENZ-45012-0010
Methylated DNA IP kit, BIOARRAY™
Catalog # ALEXENZ-45012-0010
Supplier:  
CAS Number:  
Methylated DNA IP kit, BIOARRAY™
BIOARRAY™ Methylated DNA IP kit, 10 Tests
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BIOARRAY METHYLATED DNA IP KIT 1 * 10 Tests Catalog #ENZOENZ450120010
Product ENZOENZ450120010 is a replacement for ALEXENZ-45012-0010 and has the same packaging and specifications, but may be a different quantity.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
BIOARRAY™ Methylated DNA IP Kit features IP technology for the enrichment of 5-methylcytosine-containing DNA from any pool of fragmented genomic DNA for use in genome-wide methylation analysis in the completion of diverse downstream applications.
  • Highly specific antibody is included to distinguish methylated DNA from pool
  • DNA enrichment for large-scale DNA methylation analysis
  • Easy to use with reproducible results
The kit includes a highly specific anti-5-methylcytosine monoclonal antibody for the capture and separation of methylated DNA from non-methylated DNA in only a few hours.
 : Kit contains Protein A Beads, Anti-5 methylcytosine, Control DNA, Control primer 1 , Control primer 2, IP Buffer, DNA denaturing Buffer, Control mouse DNA (methylated/nonmethylated), DNA elution buffer, magnetic rods.
Specifications
Specifications
  • Pk:
    10 Tests
  • Description:
    BIOARRAY™ Methylated DNA IP kit