Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
 

Cell lysis buffer, RIPA

RIPA cell lysis buffer 3, 7,4, 100 ml
Catalog # ASSAADI-80-1496
Cell lysis buffer, RIPA
Catalog # ASSAADI-80-1496
Supplier:  
CAS Number:  
Cell lysis buffer, RIPA

RIPA cell lysis buffer 3, 7,4, 100 ml

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
RIPA CELL LYSIS BUFFER 3, (100 ML) 1 * 100 mL Catalog #ENZOADI801496

Product ENZOADI801496 is a replacement for ASSAADI-80-1496 and has the same packaging and specifications, but may be a different quantity.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

The RIPA liquid cell lysis buffer composes of 50 mM TRIS-hydrogen chloride, pH 7,4, containing 150 mM sodium chloride, 1 mM EDTA, 1 mM EGTA, 1,2% Triton X-100, 0,5% sodium deoxycholate, and 0,1% SDS.

Storage temperature: 4 °C

Caution: RUO - Research use only.

Specifications

Specifications

  • Pk:
    100 ml
  • Description:
    RIPA cell lysis buffer 3
  • pH value:
    7,4
  • Cat. No.:
    ASSAADI-80-1496
  • Formulation:
    Liquid. 50 mM Tris, containing 150 mM NaCl, 1 mM EDTA, 1 mM EGTA, 1,2% Tergitol 15-S-9, 0,5% sodium deoxycholate, and 0,1% SDS
  • Storage temperature:
    4 °C (Long term storage)
  • Shipping conditions:
    20

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.