Analytical Guard Column, HxSil C8, Reversed Phase, 10µm, 2.1mm * 33mm, Stainless Steel 1 * 1 items
HAMI79298
Analytical Guard Column, HxSil C8, Reversed Phase, 10µm, 2.1mm * 33mm, Stainless Steel 1 * 1 items
HAMI79298
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection